Learn More
Invitrogen™ BLOCK-iT™ Inducible Pol II miR RNAi Expression Vector Kit with EmGFP
Product Code. 10688713
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10688713 20 Reactions 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10688713 Supplier Invitrogen™ Supplier No. K493900
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The BLOCK-iT™ Pol II miR RNAi Expression Vector Kits combine the advantages of traditional RNAi vectors (stable expression and the ability to use viral delivery) with capabilities for tissue-specific expression and multiple target knockdown from the same transcript.
The BLOCK-iT™ Pol II miR RNAi Expression Vector Kits combine the advantages of traditional RNAi vectors (stable expression and the ability to use viral delivery) with capabilities for tissue-specific expression and multiple target knockdown from the same transcript. The BLOCK-iT™ Pol II miR RNAi Expression Vector Kits and the BLOCK-iT™ Lentiviral Pol II miR RNAi Expression Systems are designed to express artificial miRNAs which are engineered to have 100% homology to your target sequence and will result in target cleavage. Using Invitrogen's award-winning BLOCK-iT™ RNAi Designer, over 70% of constructs produce more than 70% knockdown.
The Invitrogen Pol II expression family of vectors include:
  • Strong expression from the CMV immediate early promoter, with the option to use tissue-specific or other regulated promoters via MultiSite Gateway™ recombination.
  • Compatibility with many of Invitrogen's Gateway™ destination (DEST) vectors for gene expression.
  • Ability to control the initiation of the RNAi response with a new BLOCK-iT™ Inducible Pol II miR RNAi Expression Vector Kit w/EmGFP.
  • A new destination vector in the BLOCK-iT™ HiPerform™ Lentiviral PolII miR RNAi Expression System with EmGFP. The new vector contains an mRNA stabilizing sequence (WPRE) and a nuclear import sequence (cPPT) which generate up to 5-fold higher virus titers and EmGFP expression levels. Additionally, blasticidin resistance is expressed from the mouse Pgk-1 promoter to avoid shut-down after multiple passages.
  • Co-cistronic expression of Emerald GFP (EmGFP), resulting in correlation of EmGFP expression with knockdown from your miR RNAi.
  • Expression of more than one engineered miR RNAi sequence on the same transcript, allowing the knockdown of multiple genes simultaneously and the generation of synthetic phenotypes
The new BLOCK-iT™ Inducible Pol II miR RNAi Expression Vector Kit with EmGFP provides the ability to regulate RNAi experiments. This kit contains the pT-REx-DEST30 Gateway™ vector which after simple cloning and shuttling techniques, produces a miR RNAi expression vector suitable for inducible knockdown. The pT-REx-DEST30 Gateway™ vector contains the CMV promoter with two copies of the tetracycline operator (tetO2) sequence allowing high-level and regulated expression. This permits the study of loss of function in a stably transfected cell line even if the gene of interest is essential. Also, induction of miR RNAi expression can be halted so phenotypic changes can be measured during recovery of gene function.
For a variety of expression options, the miR RNAi cassette, which contains EmGFP (pcDNA™6.2-GW/EmGFP-miR vector only), miR flanking regions, and an miRNA homologous to the target of interest, can be readily moved into a variety of DEST vectors. This occurs through Gateway™ recombination reactions in which the miR RNAi cassette is transferred into a pDONR™ vector (BP reaction) and then into a DEST vector (LR reaction) of choice. The HiPerform™ system has the new pLenti6.4/RF R2/V5-DEST destination vector for high virus titers and EmGFP expression.
TRUSTED_SUSTAINABILITY
Specifications
Product Type RNAi Expression Vector Kit, BLOCK-iT™ Inducible Pol II miR RNAi Expression Vector Kit with EmGFP
Description BLOCK-iT™ Inducible Pol II miR RNAi Expression Vector Kit with EmGFP, 20rxns, Liquid, CMV/TO Promorter, Transfection Delivery Method, Gateway™ Cloning Method, BLOCK-iT RNAi Vectors Type, Inducible System, GFP (EmGFP) Reporter,
For Use With (Application) RNAi, epigenetics and non-coding RNA research, vector-based RNAi
No. of Reactions 20
Vector BLOCK-iT RNAi vectors
Format Kit
Gene GFP (EmGFP)
Content And Storage BLOCK-iT™ Inducible PolII miR RNAi Expression Vector Kit w/EmGFP
A cloning box with:
• Linearized pcDNA™6.2-GW/EmGFP-miR
• Annealing buffer (10X)
• T4 DNA Ligase
• DNA Ligation buffer (5X)
• lacZ control oligo
• lacZ control plasmid
• Negative control plasmid
• DNase/RNase-free water
• Forward and reverse sequencing primers
pT-REx-DEST30 Gateway™ Vector Kit
• pT-REx-DEST30
• pT-REx/GW-30/lacZ vectors
T-REx™ Regulatory Module
• pcDNA™6/TR
• Tetracycline
The Gateway™ LR and BP Clonase™ kits, blasticidin, and pDONR™ 221 are also included.
Store the vectors, buffers, control oligo and plasmids, antibiotics, clonase kits, water, and sequencing primers at -20°C. The One Shot™ box contains transformation reagents including twenty-one 50-μL aliquots of One Shot™ TOP10 Chemically Competent E. coli, S.O.C. medium, and a pUC19 supercoiled control plasmid. Store these transformation reagents -80°C.
Cloning Method Gateway™
Constitutive or Inducible System Inducible
Delivery Type Transfection
RNAi Type miRNA
Selection Agent (Eukaryotic) Blasticidin
Promoter CMV/TO
Product Line BLOCK-iT
Quantity 20 Reactions
Show More Show Less
For Research Use Only. Not for use in diagnostic procedures.