Learn More
BD Safety-Lok™ Blood Collection Set with 30 cm Tubing
Product Code. 15856161
Change view
Click to view available options
Needle Gauge:
21 G
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Unit Size
15856161 21 G Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15856161 Supplier BD Supplier No. 367286
  • ONLINE EXCLUSIVE PRICE
    243.16€
    199.76€ / 200 pièces
    Économisez 43.40€
    17% de réduction
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Use for blood collection. BD Vacutainer™ Safety-Lok™ Blood Collection Set with 30 cm tubing comes with safety re-shielding cover and Luer adapter.
  • Includes safety re-shielding cover and Luer adapter,
  • 21 G ¾ in., 30 cm tubing, green
  • Recommended for adults
  • One case consists fo 4 x 50 Blood Collection Sets
  • One Set is necessary for a pair of blood culture bottles
TRUSTED_SUSTAINABILITY
Specifications
Product Type Blood Collection Set
Color Green
Needle Gauge 21 G
Includes With luer adapter, 21 guage, 19mm needle size, 305mm tubing
Length (Metric) Needle 19 mm
Length (Metric) Tube 305 mm
Quantity 200 Cs.