Learn More
Grant Instruments™ Test Tube Rack for Unstirred Water Baths
Product Code. 11436397
Change view
Click to view available options
Holds:
105 0.5 mL Tubes
12 30 mm Tubes
18 25 mm Tubes
32 19 mm Tubes
36 16 mm Tubes
55 13 mm Tubes
65 1.5 mL Tubes
84 10 mm Tubes
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
11436397 32 19 mm Tubes Each
11466397 105 0.5 mL Tubes Each
11456397 12 30 mm Tubes Each
11476397 65 1.5 mL Tubes Each
11416397 55 13 mm Tubes Each
11426397 36 16 mm Tubes Each
11446397 18 25 mm Tubes Each
11406397 84 10 mm Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 11436397 Supplier Grant Instruments™ Supplier No. J219
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Keeps test tubes upright and secure in the bath.
  • Compatible with a wide range of Grant Unstirred Water Baths
  • Holds up to 105 test tubes depending on size
  • Dual slotted trays keep tubes secure and prevent tipping
TRUSTED_SUSTAINABILITY
Specifications
Holds 32 19 mm Tubes