Learn More
Kimble Chase™ Borosilicate Glass Chromatography Column with Reservoir, Standard Taper Joint and Fritted Disc
Product Code. 11872943 Shop All DWK Life Sciences Products
Change view
Click to view available options
Length (Metric):
250 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11872943 250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11872943 Supplier Kimble Chase™ Supplier No. 4206200425
  • ONLINE EXCLUSIVE PRICE
    269.85€
    222.62€ / 1 pièces
    Économisez 47.23€
    17% de réduction
 Restricted Item
Expédition estimée: 17-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Quantity 1 Each
Diameter (Metric) Inner 38 mm
Type Chromatography Column
USP Type Type I
Column Capacity 280 mL
Length (Metric) 250 mm
Material Borosilicate Glass
Includes With reservoir, standard taper joint and fritted disc.