Learn More
BUCHI Condensers for BUCHI Rotavapors
Product Code. 11977198 Shop All Buchi Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
Rotavapor R-100
Rotavapor R-210/R-215
Rotavapor R-210/R-215 R-210/R-215
Material (Coating):
None
P+G
Type:
Condenser C
Condenser S
Diagonal Condenser
Vertical Condenser V
missing translation for 'unitSize':
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Material (Coating) Type missing translation for 'unitSize'
11977198 Rotavapor R-210/R-215 R-210/R-215 P+G Diagonal Condenser Each
10215222 Rotavapor R-210/R-215 None Condenser C Each
10077051 Rotavapor R-210/R-215 None Condenser S Each
10302282 Rotavapor R-210/R-215 P+G Condenser S Each
12763407 Rotavapor R-100 None Vertical Condenser V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11977198 missing translation for 'mfr' BUCHI missing translation for 'supplierNo' 047976
Expédition estimée: 25-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Original BUCHI glassware guarantees the highest levels of efficiency and safety thanks to high precision in manufacturing. For more than 80 years, BUCHI has been manufacturing innovative and durable glass parts to ease daily laboratory work.
  • High throughput due to advanced design of condensers
  • Highly resistant glass provides the highest levels of safety (DURAN™ borosilicate glass 3.3)
  • Guaranteed leak-tightness and protection against hazardous fumes thanks to high precision joints
  • Maximum stability and safety with plastic and glass coating
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
Type Diagonal Condenser
For Use With (Application) Evaporator Condensation
For Use With (Equipment) Rotavapor R-210/R-215 R-210/R-215
Material (Coating) P+G
Quantity 1 Ea.