Learn More
Buffer, Reference Standard, pH 3.00 ± 0.01 at 25°C, Ricca Chemical
Product Code. 31749476
Change view
Click to view available options
Quantity:
20 x 20 mL
Packaging:
Pack
Unit Size:
20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31749476 20 x 20 mL Pack 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31749476 Supplier Ricca Chemical Company Supplier No. R149500020PK
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Chemical Name or Material Buffer, Reference Standard, pH 3.00 ± 0.01 at 25°C
CAS 877-24-7, 7647-01-0, 7732-18-5
Melting Point 0°C
Boiling Point 100°C
CAS Min % 98.9, 1.02, 0.08
CAS Max % 98.9, 1.02, 0.08
Color Colorless
Density 1
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 3
Physical Form Liquid
Quantity 20 x 20 mL
Solution Type pH Buffer
Packaging Pack
Solubility Information Miscible in water
Grade Not Available