Learn More
BW B70C, TRC
Product Code. 30333315
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30333315 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333315 Supplier Toronto Research Chemicals Supplier No. TRCB820010250MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-[(E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea
CAS 134470-38-5
Molecular Formula C17 H17 F N2 O3
Quantity 250 mg
Synonym BW B70C, N-[3-[3-(4-Fluorophenoxy)phenyl]-1-methyl-2-propen-1-yl]-N-hydroxyurea, BW-B 70C, N-[3-[3-(4-fluorophenoxy)phenyl]-1-methyl-2-propenyl]-N-hydroxy-urea
InChI Formula InChI=1S/C17H17FN2O3/c1-12(20(22)17(19)21)5-6-13-3-2-4-16(11-13)23-15-9-7-14(18)8-10-15/h2-12,22H,1H3,(H2,19,21)/b6-5+
SMILES CC(\C=C\c1cccc(Oc2ccc(F)cc2)c1)N(O)C(=O)N
IUPAC Name 1-[(E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea
Molecular Weight (g/mol) 316.327
Formula Weight 316.122