Learn More
Gibco™ C57BL/6 Mouse Embryonic Fibroblasts, MitC-treated
Product Code. 15960453 Shop All Gibco Products
Change view
Click to view available options
Quantity:
4 x 106 cells
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15960453 4 x 106 cells Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15960453 Supplier Gibco™ Supplier No. A34962
Cell Culture Essentials
Use promo code 27976
View all promotions
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Mitomycin C (MitC)-treated Gibco C57BL/6 Mouse Embryonic Fibroblasts (MEFs) are feeder cells that support the culture of healthy undifferentiated embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs), especially mouse ESCs/iPSCs.
Manufactured by MTI-GlobalStem and equivalent to MTI-GlobalStem Cat. No. GSC-6002M, these MEF cells are isolated from inbred C57BL/6 mice, mitotically arrested by MitC treatment, and tested to help ensure performance and minimize the risk of contamination. For our most widely used feeder cells for culturing pluripotent stem cells, see CF1 Mouse Embryonic Fibroblasts, Irradiated or CF1 Mouse Embryonic Fibroblasts, MitC-Treated. You may also choose from our selection of drug-resistant MEFs to suit your application.
• Help save precious time with pre-inactivated, ready-to-use MEFs
• Culture ESCs and iPSCs with confidence using meticulously tested feeder cells
• Provide optimum support for mouse ESCs and iPSCs
Help save time with ready-to-use MEFs
MitC-treated C57BL/6 MEFs help save time and trouble dealing with an animal facility, performing tedious, time-consuming dissections, and expanding and testing cells day after day, week after week. Focus on your research with these pre-inactivated, ready-to-use feeders.
Culture with meticulously tested feeder cells
Each lot of MitC-treated C57BL/6 MEFs is tested for bacteria, fungi, mycoplasma, and mouse pathogens to minimize the risk of contamination. Each lot is also tested for post-thaw viability (typically greater than 95%) and ability to support mouse and human ESCs to help ensure robust and consistent performance. Culture your ESCs and iPSCs with confidence, knowing that you can rely on these high quality feeder cells.
Provide optimum support for mouse ESCs and iPSCs
While C57BL/6 MEFs can also support human ESCs and iPSCs, these feeder cells work particularly well for culturing mouse ESCs and iPSCs, showing better results compared to CF1 and ICR strain MEFs.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Cryopreserved mouse embryonic fibroblasts
Store in liquid nitrogen.
Age Embryonic
Cell Type Feeder Cells (Mouse Embryonic Fibroblast)
Form Cryopreserved
Species Mouse
Donor Source Pooled
No. of Cells 4 x 106
Quantity 4 x 106 cells
Shipping Condition Shipped on dry ice
For Research Use Only. Not for use in diagnostic procedures.