Learn More
Calcium (2R,3S)-2,3,4-trihydroxybutanoate, 95%, BLD Pharm™
Product Code. 31428088
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31428088 100 g
31419037 25 g
31426694 5 g
31415412 1 kg
31416065 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31428088 Supplier BLD Pharm Supplier No. BD136793100g
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 70753-61-6
Molecular Formula C8H14CaO10
MDL Number MFCD00077882
Quantity 100 g
SMILES O=C([O-])[C@H](O)[C@@H](O)CO.O=C([O-])[C@H](O)[C@@H](O)CO.[Ca+2]
Molecular Weight (g/mol) 310.27
Percent Purity 95%
Chemical Name or Material Calcium (2R,3S)-2,3,4-trihydroxybutanoate