Learn More
Calcium Pantothenate, U.S.P., J.T. Baker™
Product Code. 15517914 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15517914 1 kg Wide Mouth Amber Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15517914 Supplier Avantor J.T.Baker Supplier No. 144308
  • ONLINE EXCLUSIVE PRICE
    3471.90€
    2384.61€ / 1kg
    Économisez 1087.29€
    31% de réduction
 Restricted Item
Expédition estimée: 20-05-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Chemical Identifiers
CAS 137-08-6
Molecular Formula C18H32CaN2O10
Molecular Weight (g/mol) 476.54
MDL Number MFCD00002766
InChI Key FAPWYRCQGJNNSJ-DXHDTSSINA-L
Synonym calcium pantothenate, d-pantothenic acid hemicalcium salt, d-pantothenic acid, calcium salt, pantothenic acid, calcium salt, d, r-n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-beta-alanine calcium salt, beta-alanine, n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-, calcium salt, r
PubChem CID 131847364
IUPAC Name calcium bis(3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate)
SMILES [Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O
Specifications
Quantity 1 kg
Packaging Wide Mouth Amber Glass Bottle
Formula Weight 476.54
Percent Purity 98.8%
Grade USP
Chemical Name or Material Calcium Pantothenate