Learn More
Dueperthal™ Can Collector
Product Code. 10546512
Change view
Click to view available options
Capacity (Metric):
12 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10546512 12 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10546512 Supplier Dueperthal™ Supplier No. 5.10.010
 Restricted Item
Expédition estimée: 22-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Capacity (Metric) 12 L
Diameter (Metric) Outer 250 mm
Color Blue
Finish Stove enamelled
Height (Metric) 350 mm
For Use With (Application) Use as safety containers or cleaning containers