Learn More
Thermo Scientific Chemicals Caspase-7 Substrate I, fluorogenic
Product Code. 15497899
Change view
Click to view available options
Substrate Quantity:
0.5mg
1mg
Unit Size:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Substrate Quantity Unit Size
15497899 0.5mg 0.5mg
15407909 1mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497899 Supplier Thermo Scientific Chemicals Supplier No. J66513.!D
 Restricted Item
Expédition estimée: 15-12-2026
Add to basket
This item is not returnable. View return policy
For cellular and molecular biology applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Color White to off-white
Form Lyophilized
Substrate Type Caspase Substrate
Molecular Weight (g/mol) 1327.31
Storage Requirements -30° to -10°C
Substrate Quantity 0.5mg