Learn More
TPP™ Cell Scrapers
Product Code. 11301724
Change view
Click to view available options
Width (Metric) Blade:
20 mm
25 mm
Unit Size:
Pack of 100
Pack of 150
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Blade Unit Size
11301724 25 mm Pack of 100
11391714 20 mm Pack of 150
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11301724 Supplier TPP™ Supplier No. 99004
  • ONLINE EXCLUSIVE PRICE
    464.00€
    387.10€ / 100 pièces
    Économisez 76.90€
    16% de réduction
Add to basket
Edge
Add to basket
TPP™ cell scraper require slight pressure on handle using forefinger, thus pushing handle downward towards floor of container.
    Free rotating blade
  • Total access to all corner
  • Different blade widths
  • Small raised knobs on handle
  • Lint-free blister packaging
  • User friendly TPP dispenser case
Specifications
Width (Metric) Blade 25 mm
For Use With (Equipment) Tissue culture flask T-300
No. per Case 100/Cs.
Product Type Cell Scraper
Material Polyethylene