Learn More
Invitrogen™ Celleste™ 3D Deconvolution Module, network license (includes one seat)
Code produit 16688977 Tous les produits Thermo Scientific Produits
Modifier l'affichage
Click to view available options
Quantity:
1 each
Conditionnement:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
16688977 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16688977 Fournisseur Invitrogen™ Code fournisseur AMEP4916
Expédition estimée: 07-12-2026
The Celleste 5.0 Image Analysis package easily transforms images into quantitative data in a streamlined workflow. The Celleste 3D Deconvolution Module for Network License helps dramatically improve resolution and clarity of thick samples like spheroids, tissue slices, or cells in 3D matrices by deconvolving image Z-stacks.
The Celleste 5.0 Image Analysis package easily transforms images into quantitative data in a streamlined workflow. The Celleste 3D Deconvolution Module for Network License helps to dramatically improve resolution and clarity of thick samples like spheroids, tissue slices, or cells in 3D matrices by deconvolving image Z-stacks. It also helps enhance image quality for presentation, publication, or 3D volume analysis. Coupled with the powerful image acquisition capabilities of the EVOS microscopes, Celleste 5 software allows you to seamlessly capture, process, measure, analyze and share images and data. A Network Server License is required for use with this module.
Celleste 5.0 Image Analysis Software Network License [1] is a full-feature image analysis package designed for a range of biological applications, from image adjustments and processing with manual and automatic measurements over multiple channels, to segmentation and classification tools that help you transform images into quantitative data in a streamlined and customizable workflow. The Celleste 5.0 Image Analysis Network Server License is designed for environments where the use of multiple copies is desired. The Network Server License comes with one seat and additional Soft License Seats [2] can be added at any time. Optional modules for 2D and 3D deconvolution, visualization, analysis and GPU-powered processing allow you to customize the capabilities according to your needs. The Celleste 3D Deconvolution Module for Network License helps to dramatically improve resolution and clarity of thick samples like spheroids, tissue slices, or cells in 3D matrices by deconvolving image Z-stacks. It also helps enhance image quality for presentation, publication, or 3D volume analysis. Coupled with the powerful image acquisition capabilities of the EVOS microscopes [3], Celleste 5 software allows you to seamlessly capture, process, measure, analyze and share images and data.Note that Celleste 5.0 Image Analysis Network Server License is required for use with any of the modules.Celleste 5.0 software with the Celleste 3D Deconvolution Module for Network License offers you these important advantages:• Streamlined workflow tools that make it easy to visualize and edit images• Powerful image analysis capabilities for segmentation, classification, and quantification of single images or a batch of images• 3D blind and non-blind deconvolution options• Advanced PSF controls with measured and theoretical PSF options• A suite of 3D display and visualization tools• Dramatically improved image qualityMore about the deconvolution modulesDeconvolution is an algorithm-based process used to restore systematic loss of image contrast (blur) that results from the image point spread function (PSF). The powerful 2D and 3D restoration algorithms available in the Celleste 5 modules allow you to deconvolve your images to enhance contrast and quality, or to visualize them in Z, over time and by channel, then analyze the resulting image using an expanded tool set.[1] Celleste 5.0 Image Analysis Network Server License [AMEP4868][2] Celleste Image Analysis Network Seat [AMEP4869]Explore the entire EVOS line of imaging systems, accessories, and offers [3] [www.thermofisher.com/EVOS]For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Spécification
Product Line Celleste
For Use With (Equipment) EVOS M5000, EVOSM7000 and Other Imaging Systems
Quantity 1 each