Learn More
3M™ Center Adapter Assembly 6864/37000
Product Code. 15769637 Shop All 3M Products
Change view
Click to view available options
Color: Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15769637 Color: Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15769637 Supplier 3M™ Supplier No. 7000002178
  • ONLINE EXCLUSIVE PRICE
    40.00€
    25.97€ / 1 pièces
    Économisez 14.03€
    35% de réduction
Expédition estimée: 04-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
3M™ Center Adapter Assembly 6864/37000 includes center port adaptor, exhalation valve and lens gasket for the 3M™ Full Face Mask 6000 Series.
  • Replacement full face respirator center adapter assembly includes center port adaptor, exhalation valve and lens gasket
  • Replace damaged parts to ensure correct levels of protection and comfort
  • For use with the 3M™ 6000 Series Full Face Respirator
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Product Type Center Adapter Assembly
Quantity 5/Box