Learn More
Eppendorf Centrifuge 5427 R (EU-IVD)
BIOPHARMA
Product Code. 17894744 Shop All Eppendorf Products
Change view
Click to view available options
Electrical Requirements:
230V, 50/60Hz (EU)
Includes:
With Rotor FA-45-30-11
With Rotor FA-45-48-11
Without rotor
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements Includes Unit Size
17894744 230V, 50/60Hz (EU) Without rotor Each
17804754 230V, 50/60Hz (EU) With Rotor FA-45-48-11 Each
17834754 230V, 50/60Hz (EU) With Rotor FA-45-30-11 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17894744 Supplier Eppendorf Supplier No. 5429000015
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Centrifuge 5427 R contains a natural refrigerant (R290) to protect your samples and the environment. It features 9 different rotor options including an aerosol-tight 48-place rotor for high-throughput applications.
  • Rotor capacity of 48 x 1.5/2.0 mL and 12 x 5.0 mL
  • QuickLock rotor for easy lid locking
  • Max. speed of 25,001 x g (16,220 rpm)
  • High-capacity 48-place rotor increases productivity and saves time
  • Soft-touch one-finger closure for ergonomic operation
  • Temperature range from -11 °C to 40 °C
  • FastTemp function for quick pre-cooling (11 minutes from ∼23°C to 4°C)
  • ECO shut-off programmable after 1, 2, 4 or 8 hours of non-use for energy savings and compressor life
  • Built-in condensation drain eliminates water accumulation
  • Automatic rotor recognition and imbalance detection for safety
TRUSTED_SUSTAINABILITY
Specifications
Capacity 48 x 1.5/2.0 mL, 12 x 5.0 mL
Max. Speed 16,220 rpm
Max. RCF 25,001 x g
Height (English) Exterior 10 in.
Height (Metric) Exterior 25.4 cm
Noise Level < 55 dBA with rotor FA-45-48-11
Timer 5 sec. to 9:59 hr.
Width (English) Exterior 12.6 in.
Width (Metric) Exterior 31.9 cm
Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
Includes Without rotor
Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, certified aerosol-tight rotor lids
Electrical Requirements 230V, 50/60Hz (EU)
Model Centrifuge 5427 R, rotary knobs
Wattage 495 W
Refrigerated Yes
Voltage 230 V
Product Type Microcentrifuge
Display Type Large, brightly lit LCD
Show More Show Less
The Centrifuge 5427 R is a non-automatic centrifuge for the separation of liquid substance mixtures from the human body and is specifically intended to be used as an accessory together with an in vitro diagnostic device and to enable its use according to its intended purpose. This product complies with the requirements of the EU In Vitro Diagnostics Regulation (IVDR) 2017/746. Eppendorf centrifuges are exclusively intended for indoor use by trained specialists.