Learn More
Eppendorf™ Centrifuge 5804/5804 R
BIOPHARMA
Product Code. 15825103 Shop All Eppendorf Products
Change view
Click to view available options
Includes:
Rotor A-4-44 incl. adapters for 15/50 mL conical tubes
Rotor S-4-72 incl. adapters for 15/50 mL conical tubes
Without rotor
Electrical Requirements:
230 V, 50/60 Hz (EU)
Model:
Centrifuge 5804
Centrifuge 5804 (EU-IVD)
Centrifuge 5804 R
Centrifuge 5804 R (EU-IVD)
Unit Size:
1 set
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Electrical Requirements Model Unit Size
15825103 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5804 R Each
10022724 Rotor A-4-44 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 (EU-IVD) Each
12806213 Rotor S-4-72 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 (EU-IVD) Each
10344701 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5804 (EU-IVD) Each
15885093 Rotor A-4-44 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 1 set
15805103 Rotor S-4-72 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 1 set
15865093 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5804 Each
10246074 Rotor A-4-44 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 R (EU-IVD) Each
12816213 Rotor S-4-72 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 R (EU-IVD) Each
10334751 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5804 R (EU-IVD) Each
15845103 Rotor A-4-44 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 R 1 set
15865103 Rotor S-4-72 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 R 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 15825103 Supplier Eppendorf™ Supplier No. 5805000010
  • ONLINE EXCLUSIVE PRICE
    8525.00€
    7767.00€ / 1 pièces
    Économisez 758.00€
    8% de réduction
View all promotions
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The high-speed Eppendorf Centrifuge 5804/5804 R is a medium capacity centrifuge, ideal for molecular applications in tubes up to 250 mL. It also offers supreme versatility with the capability of taking swing-bucket and fixed-angle rotors, as well as deepwell plates.
  • Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 mL to 250 mL
  • Plate rotor for centrifugation of all types of MTP, PCR, cell culture, or Deepwell Plates
  • Fixed-angle rotors for high-speed applications in tubes from 0.2 mL to 85 mL
  • High centrifugation speed of up to 20,913 × g (14,000 rpm)
  • Centrifuge lid with soft-touch lid closure
  • Low access height of 29 cm for easy loading and unloading of samples
  • Quiet operation to improve your work environment
  • Compact footprint saves valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
    Controller Type Keypad
    Max. RCF 20,913 x g
    Max. Speed 14,000 rpm
    Refrigerated Yes
    Includes Without rotor
    Model Centrifuge 5804 R
    Noise Level <58 dB(A)
    Rotor Included No
    Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids and aerosol-tight caps, Rotor lifecycle counter, automatic imbalance recognition
    Standards EN ISO 12100, EN 61010-1, EN 61010-2-020, EN 61326-1, EN 55011, CFR 47 FCC part 15 class A, EN 378-1, EN 378-2, EN 50581 (refer to manual for more details)
    Timer 1 to 99 min., with continuous run function, short-spin
    Dimensions (L x W x H) Exterior 64 x 55 x 34 cm
    Width (English) Exterior 25.0 in.
    Width (Metric) Exterior 63.4 cm
    Height (English) Exterior 13.5 in.
    Height (Metric) Exterior 34.2 cm
    Electrical Requirements 230 V, 50/60 Hz (EU)
    Voltage 230 V
    Wattage 1650 W
    Frequency 50/60 Hz
    Product Type Benchtop Centrifuges
    Temperature Range (Metric) -9°C to +40°C
    Display Type Large, brightly lit LCD