Learn More
Nabertherm™ Chimney
Product Code. 12201282 Shop All Nabertherm Products
Change view
Click to view available options
For Use With (Application):
Exhaust Gas Removal from Furnace
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
12201282 Exhaust Gas Removal from Furnace Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12201282 Supplier Nabertherm™ Supplier No. 6000140311
  • ONLINE EXCLUSIVE PRICE
    657.00€
    590.00€ / 1 pièces
    Économisez 67.00€
    10% de réduction
View all promotions
 Restricted Item
Expédition estimée: 15-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Chimney with fan, to remove exhaust gas from the furnace better
not suitable for
  • L 15...
  • L 1/12
  • LE 1/11
  • LE 2/12
Specifications
Controller Type P330
Type Chimney
For Use With (Application) Exhaust Gas Removal from Furnace
Includes Fan