Learn More
Chlorguanide-d4 Hydrochloride, TRC
Product Code. 30361510
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361510 10 mg 10mg
30355621 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30361510 Supplier Toronto Research Chemicals Supplier No. TRCC32950210MG
 Restricted Item
Expédition estimée: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 1189671-34-8
Molecular Formula C11H13D4Cl2N5
Molecular Weight (g/mol) 294.22
Synonym N-(4-Chlorophenyl-d4)-N'-(1-methylethyl)-imidodicarbonimidic Diamide Hydrochloride, 1-(p-Chlorophenyl-d4)-5-isopropylbiguanide Hydrochloride, Bigumal-d4, Chlorguanid-d4, Chloroguanide-d4, Paludrin-d4, Proguanil-d4
IUPAC Name 1-((l2-azaneylidene)((4-chlorophenyl-2,3,5,6-d4)amino)methyl)-3-isopropylguanidine
SMILES CC(C)NC(NC(NC1=C([2H])C([2H])=C(Cl)C([2H])=C1[2H])=N)=N.Cl
Specifica
Quantity 10 mg
InChI Formula InChI=1S/C11H16ClN5.ClH/c1-7(2)15-10(13)17-11(14)16-9-5-3-8(12)4-6-9;/h3-7H,1-2H3,(H5,13,14,15,16,17);1H/i3D,4D,5D,6D;
Formula Weight 293.11
Chemical Name or Material Chlorguanide-d4 Hydrochloride
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.