Learn More
Chlormezanone, Mikromol™
Product Code. 17901036
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17901036 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17901036 Supplier Mikromol Supplier No. MM0017.00
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Chlormezanone
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Chlormezanone
CAS 80-77-3
Analyte or Component Names Chlormezanone
Shipping Condition Room Temperature
Molecular Formula C11 H12 Cl N O3 S
Certifications/Compliance ISO 17025
Synonym Chlormezanone
InChI Formula InChI=1S/C11H12ClNO3S/c1-13-10(14)6-7-17(15,16)11(13)8-2-4-9(12)5-3-8/h2-5,11H,6-7H2,1H3
SMILES CN1C(c2ccc(Cl)cc2)S(=O)(=O)CCC1=O
IUPAC Name 2-(4-chlorophenyl)-3-methyl-1,1-dioxo-1,3-thiazinan-4-one
Molecular Weight (g/mol) 273.74
Quantity 250 mg
Formula Weight 273.0226 g/mol