Learn More
Chloropentafluorobenzene, 98%, BLD Pharm™
Product Code. 30965623
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30965623 500 g 500g
30957200 25 g 25g
30962751 5 g 5g
30952017 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30965623 Supplier BLD Pharm Supplier No. BD120914500g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 344-07-0
Molecular Formula C6ClF5
MDL Number MFCD00000534
Quantity 500 g
SMILES FC1=C(Cl)C(F)=C(F)C(F)=C1F
Molecular Weight (g/mol) 202.51
Percent Purity 98%
Chemical Name or Material Chloropentafluorobenzene