Learn More
Macherey-Nagel™ Chromafix™ PS-OH- cartridges
Product Code. 10796132 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
50 Pack
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10796132 50 Pack Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10796132 Supplier Macherey-Nagel™ Supplier No. 731868
  • ONLINE EXCLUSIVE PRICE
    265.00€
    243.60€ / 50 pièces
    Économisez 21.40€
    8% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Base Material Strong PS/DVB Cation Exchanger, OH- form
For Use With (Application) Removal or Concentration of Anions From Water Increasing The pH Value in Acidic Samples
Particle Size 100 μm
Type Chromafix PS-OH- Cartridge
Pore Size 100 Å
Quantity 50 Pack