missing translation for 'onlineSavingsMsg'
Learn More

chroman-8-carbonyl chloride, Thermo Scientific™

Product Code. 10265814
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265814 5 g Amber Glass Bottle 5g
10653455 250 mg Amber Glass Bottle 250mg
10505384 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265814 Supplier Thermo Scientific Maybridge Supplier No. CC76102DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1034566-09-0
Molecular Formula C10H9ClO2
Molecular Weight (g/mol) 196.63
InChI Key TTYLUAYKWAEQDM-UHFFFAOYSA-N
Synonym chroman-8-carbonyl chloride, 3,4-dihydro-2h-1-benzopyran-8-carbonyl chloride, chromane-8-carbonyl chloride
PubChem CID 15480465

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 196.63
Percent Purity 90%
Chemical Name or Material chroman-8-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.