Learn More
Chromone-3-carbonitrile, 97%, Thermo Scientific™
Product Code. 11447637
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447637 5 g 5g
11437637 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447637 Supplier Thermo Scientific Alfa Aesar Supplier No. A15687.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 50743-17-4
Molecular Formula C10H5NO2
Molecular Weight (g/mol) 171.16
MDL Number MFCD00052604
InChI Key SFWNPLLGXKJESA-UHFFFAOYSA-N
Synonym 3-cyanochromone, chromone-3-carbonitrile, 4-oxo-4h-chromene-3-carbonitrile, 4-oxo-4h-1-benzopyran-3-carbonitrile, acmc-20an5h, maybridge1_006148, 4-oxo-chromene-3-carbonitrile, 3-cyano-4h-1-benzopyran-4-one, 4-oxo-1-benzopyran-3-carbonitrile
PubChem CID 521298
SMILES O=C1C(=COC2=CC=CC=C12)C#N
Spezifikation
Melting Point 173°C to 177°C
Quantity 5 g
UN Number UN3439
IUPAC Name 4-oxochromene-3-carbonitrile
Formula Weight 171.16
Percent Purity 97%
Chemical Name or Material Chromone-3-carbonitrile
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.