Learn More
Chromone-3-carboxylic acid, 95%, BLD Pharm™
Product Code. 30982316
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30982316 1 g -
30971221 250 mg -
30980090 5 g -
30971597 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30982316 Supplier BLD Pharm Supplier No. BD125621g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 39079-62-4
Molecular Formula C10H6O4
MDL Number MFCD00017338
Quantity 1 g
SMILES O=C1C(C(O)=O)=COC2=C1C=CC=C2
Molecular Weight (g/mol) 190.15
Percent Purity 95%
Chemical Name or Material Chromone-3-carboxylic acid