Learn More
Proteintech ChromoTek GFP-Trap™ Magnetic Agarose
Product Code. 17313363
Change view
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For Unit Size
17313363 10 Reactions 10 reactions
17323363 100 Reactions 100 reactions
17333363 20 Reactions 20 reactions
17343363 200 Reactions 200 reactions
17353363 400 Reactions 400 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17313363 Supplier Proteintech Supplier No. gtma10
  • ONLINE EXCLUSIVE PRICE
    259.27€
    237.00€ / 10 réactions
    Économisez 22.27€
    8% de réduction
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
The ChromoTek GFP-Trap™ Magnetic Agarose are affinity beads for IP of GFP-fusion proteins.
  • Comprises a GFP Nanobody/ VHH coupled to magnetic agarose beads
Specifications
Content And Storage Upon receipt store at 4°C. Do not freeze.
For Use With (Application) IP, CoIP, ChIP, RIP
Target AcGFP, Clover, eGFP, Emerald, GFP, GFP5, GFP Envy, GFP S65T, mGFP, mPhluorin, PA-GFP, Superfolder GFP, TagGFP, TagGFP2, monomeric eGFP A206K, CFP, YFP, Citrine, eCitrine, eYFP, Venus, Ypet, BFP
Label or Dye Magnetic Agarose
Species Alpaca
Sufficient For 10 Reactions
Binding Capacity 10 μL Slurry bind more than 8 μg of Recombinant GFP