Learn More
Proteintech ChromoTek pSpot8 Vector
Product Code. 17333333
Change view
Click to view available options
Quantity:
1.25 μg
Unit Size:
1.25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17333333 1.25 μg 1.25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17333333 Supplier Proteintech Supplier No. EV8
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
pSpot8 vector, S.cerevisiae, Spot C-term, Leu/Amp (Ori: 2 μ/pBR322), high expression.
  • The plasmid pSpot8 is an expression vector for Spot-Tag™ fusion proteins in S. cerevisiae. After cloning the protein of interest (POI) into the multiple cloning site (MCS) provided by pSpot8, the Spot-Tag™ (sequence: PDRVRAVSHWSS) will be fused to the C-terminus of the POI.
Specifications
Quantity 1.25 μg