Learn More
Ciclopirox Olamine, LoGiCal
Product Code. 30460753
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460753 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460753 Supplier LoGiCal Supplier No. LGCFOR0418.00
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ciclopirox olamine
CAS 41621-49-2
Analyte or Component Names Ciclopirox olamine
Shipping Condition Room Temperature
Molecular Formula C12 H17 N O2 . C2 H7 N O
Certifications/Compliance ISO 9001
Synonym Ciclopirox Olamine, 6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridinone compd. with 2-aminoethanol (1:1), 2-Aminoethanol compd. with 6-cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridinone (1:1), 1-Hydroxy-4-methyl-6-cyclohexyl-2-pyridinone ethanolamine salt, Batrafen, Brumixol, Ciclochem, Ciclopirox olamine, HOE 296, Loprox, Micoxolamina, Mycoster, NSC 336278, Terit, Ethanol, 2-amino-, compd. with 6-cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridinone (1:1) (9CI), Ciclopirox ethanolamine (1:1), Fungowas, Sebiprox, Selergo
InChI Formula InChI=1S/C12H17NO2.C2H7NO/c1-9-7-11(13(15)12(14)8-9)10-5-3-2-4-6-10;3-1-2-4/h7-8,10,15H,2-6H2,1H3;4H,1-3H2
Solution Type Neat
SMILES CC1=CC(=O)N(O)C(=C1)C2CCCCC2.NCCO
IUPAC Name 2-aminoethanol;6-cyclohexyl-1-hydroxy-4-methylpyridin-2-one
Industry Type Clinical Testing
Molecular Weight (g/mol) 268.35
Quantity 10 mg
Formula Weight 268.1787 g/mol