Learn More
Cimiracemoside D, MedChemExpress
Product Code. 16431588
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431588 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16431588 Supplier Medchem Express Supplier No. HYN0900
 Restricted Item
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Cimiracemoside D is a natural product found in Actaea racemosa with unknown details.
Cimiracemoside D is a natural product found in Actaea racemosa with unknown details.
Specifications
Chemical Name or Material Cimiracemoside D
CAS 290821-39-5
Color White
Purity Grade Notes Research
Molecular Formula C37H58O11
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (147.31 mM; Need ultrasonic)
SMILES C[C@]([C@]([C@H](OC(C)=O)C1)([C@]2([H])C3(O[C@@H]4C(C)(O)C)O[C@]4([H])C[C@H]2C)C)([C@H]3O)[C@@](CC[C@@]5([H])C6(C)C)([H])[C@@]71[C@]5(CC[C@@H]6O[C@@](OC[C@H](O)[C@@H]8O)([H])[C@@H]8O)C7
Molecular Weight (g/mol) 678.85
Formula Weight 678.85
Percent Purity 98.26%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation