Learn More
Phenomenex™ Claricep™ Acidic Alumina for Flash Columns, Irregular Media
Product Code. 31746558 Shop All Phenomenex Products
Change view
Click to view available options
Bed Weight (Metric):
100 g
1 kg
5 kg
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight (Metric) Unit Size
31746558 5 kg Each
31742087 1 kg Each
31744173 100 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31746558 Supplier Phenomenex™ Supplier No. FCA1405000A
 Restricted Item
Expédition estimée: 01-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Available in two flash media (irregular and spherical) with multiple chemistries, Claricep columns are optimized for resolution and high loading capacity. The deactivated silica surface promotes compound stability, excellent peak shape and high performance for both acidic and basic compounds.
  • Brand: Claricep
  • Technique: FLASH
  • Phase: Alumina
  • Particle Type: Irregular
TRUSTED_SUSTAINABILITY
Specifications
Type Flash Column
Bed Weight (Metric) 5 kg