Learn More
Scotch-Brite™ Clean & Shine Floor Pad
Product Code. 17198760
Change view
Click to view available options
Diameter (Metric):
280 mm
330 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
Unit Size:
Pack of 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
17198760 480 mm Pack of 5
17148770 280 mm Pack of 5
17168770 330 mm Pack of 5
17158770 355 mm Pack of 5
17188760 380 mm Pack of 5
17138770 406 mm Pack of 5
17118770 432 mm Pack of 5
17128770 460 mm Pack of 5
17108770 505 mm Pack of 5
17178770 530 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 17198760 Supplier Scotch-Brite™ Supplier No. 7100148014
  • ONLINE EXCLUSIVE PRICE
    331.00€
    196.69€ / 5 pièces
    Économisez 134.31€
    40% de réduction
Expédition estimée: 05-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
This 2 in 1 daily cleaning pad for low speed scrubbers gradually increases shine with repeated use
  • Cleans and shines in the same step, using only a low-speed scrubber
  • Save costs and labor by reducing or eliminating the need to burnish
  • Removes black marks 3 times faster than traditional cleaning pads
  • Two-sided pad lasts longer than traditional cleaning pads
  • Effective on most coated and uncoated hard floors including VCT, vinyl, rubber, marble, stone, terrazzo, granite, and concrete with only water or neutral cleaner
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleaning Pads
Color Brown
Diameter (Metric) 480 mm
For Use With (Application) Cleaning and Polishing Floors
For Use With (Equipment) Concrete, Granite, Industrial Floors, Linoleum, Marble, Stoneware, Terrazzo, VCT, and Vinyl Flooring
Style Round