Learn More
Micronova™ Cleanroom NovaSqueegee™
Product Code. 18803103 Shop All Micronova Products
Change view
Click to view available options
Product Type:
Squeegee
Squeegee Blade
Material:
Electro-polished Stainless Steel (304)
Polyvinyl Acetate
Rubber
Length (English):
12 in.
18 in.
22 in.
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Material Length (English) Unit Size
18803103 Squeegee Blade Rubber 12 in. -
17813668 Squeegee Electro-polished Stainless Steel (304) 12 in. Each
12664873 Squeegee Electro-polished Stainless Steel (304) 18 in. Each
12663506 Squeegee Electro-polished Stainless Steel (304) 22 in. Each
13107606 Squeegee Blade Polyvinyl Acetate 18 in. Each
13117606 Squeegee Blade Polyvinyl Acetate 22 in. Each
13107656 Squeegee Blade Rubber 12 in. Each
13117656 Squeegee Blade Rubber 18 in. Each
13127656 Squeegee Blade Rubber 22 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 18803103 Supplier Micronova™ Supplier No. RS12
Expédition estimée: 29-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A stainless steel squeegee tool with a replaceable rubber blade, designed to clean windows, screens, floors and walls in the cleanroom. Optional use with the SSU series universal handles allows extended reach.
The Micronova Cleanroom NovaSqueegee™ is a stainless steel tool that comes standard with a replaceable rubber blade and features a Quick Disconnect handle attachment for optional use with the SSU series handles.
  • Stainless steel squeegee tool with a rubber blade
  • Designed to clean windows, screens, floors and walls in the cleanroom
  • Made with 304 electropolished stainless steel
  • Rubber replacement blades available
  • Available in 12, 18 and 22-inch (30, 45, 55 cm) sizes
  • Quick disconnect handle attachment
  • Optional use with SSU series Universal Handles
  • Entire unit is autoclavable
Specifications
Product Type Squeegee Blade
Color-coded No
Handle Type None
Sterility Non-sterile
Autoclavable Autoclavable
Color Black
Disposable Reusable
For Use With (Application) For Cleaning Windows
For Use With (Equipment) NovaSqueegee
Length (English) 12 in.
Length (Metric) 30.5 cm
Material Rubber
Quantity 12/Cs.
Width (English) 1 in.
Width (Metric) 2.5 cm