Learn More
Thermo Scientific™ Clear C-Shaped Immuno Nonsterile 96-Well Plates
BIOPHARMA
Product Code. 10777621
Change view
Click to view available options
Surface Treatment:
MaxiSorp
PolySorp
Certified:
No
Yes
Unit Size:
Pack of 60
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Surface Treatment Certified Unit Size
10777621 MaxiSorp Yes Pack of 60
10098860 MaxiSorp No Pack of 60
10737931 PolySorp No Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Each lot is functionally tested for IgG binding.
Product Code. 10777621 Supplier Thermo Scientific™ Supplier No. 446612
  • ONLINE EXCLUSIVE PRICE
    263.00€
    218.99€ / 60 pièces
    Économisez 44.01€
    16% de réduction
View all promotions
Expédition estimée: 18-12-2026
Add to basket
Add to basket
Bulk Quote Request
Each lot is functionally tested for IgG binding.
Perform quantitative and qualitative solid phase immunoassays with these nonsterile 96-well plates, available in a variety of binding surfaces.
Perform quantitative and qualitative solid phase immunoassays with Thermo Scientific™ Clear C-Shaped Immuno Nonsterile 96-Well Plates, which include Nunc™, Immulon™ and Microtiter™ plates. Choose from a variety of binding surfaces for reliable and reproducible results in colormetric assays.
96-Well Solid Plates
  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns
Surfaces
  • PolySorp for adsorption of hydrophobic molecules
  • MaxiSorp is hydrophilic and ideal for antibody sandwich assays
Ordering Information:
Please note that availability may vary by country. 446612 includes certificate.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Binding Property Hydrophilic
Certified Yes
Format Standard
Lid Without Lid
Detection Method Colorimetric
Array 8 x 12
Packaging 60 Cases
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 250 μL
Sterility Non-sterile
Surface Treatment MaxiSorp
Well Shape Round, C Bottom
No. per Case 60
No. per Pack 5
Plate Blocking None
Target Molecule Antibodies (Glycoproteins)
Well Design C96