Learn More
Honeywell™ Clearflow 3 Compressed Airline Filtration Unit, Wall Model
Product Code. 15806435
Change view
Click to view available options
Applications:
Breathable air/air for air-driven tools
Unit Size:
1 pair
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Applications Unit Size
15806435 Breathable air/air for air-driven tools 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15806435 Supplier Honeywell™ Supplier No. 1763903
  • ONLINE EXCLUSIVE PRICE
    1263.62€
    1146.40€ / 1 paires
    Économisez 117.22€
    9% de réduction
Expédition estimée: 28-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Delivers high-quality breathable air to a range of respiratory devices.
  • Wall-mounted model with 2 CEJN outlets
  • Supplies breathable air for up to 4 workers using supplied respirators, or 2 operators and auxiliary air-driven tools
  • Uses coalescing filter and carbon absorber filter to remove oil vapor and odor
  • High filtration capacity due to high flow system, high efficiency of coalescing filter, and high capacity of absorber carbon filter
  • Easy use and easy maintenance due to new one-piece compact design
  • Provides security to worker via accurate and reliable differential pressure indicator
  • Filter
    - Coalescing filter : 1000 hours with a filtration at 99,97% to 0.01 micron - Charcoal filter : 1000 hours (or when odors are detected inside respiratory device) Filters are compliant with ISO 8573 -1 Compressed Air - Part 1: Pollutants and purity classes) which specifies the quality class of compressed air for the presence of particles, water or oil, irrespective of the mode of generation of the compressed air)
TRUSTED_SUSTAINABILITY
Specifications
Applications Breathable air/air for air-driven tools
Flow Rate 1600 L/min.
For Use With (Equipment) Respirators/air-driven tools
Material Aluminum body filters, plastic and brass regulators, polycarbonate bowl, nitrile seals, ABS cover
Pressure 8 bar