Learn More
Diversey ClearKlens ECO VH39 20L
Product Code. 17883105
Change view
Click to view available options
Volume (Metric):
20 L
Unit Size:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17883105 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17883105 Supplier Diversey Supplier No. 101107534
  • ONLINE EXCLUSIVE PRICE
    186.00€
    167.31€ / 20L
    Économisez 18.69€
    10% de réduction
 Restricted Item
Expédition estimée: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Defoamed, organic acid detergent descaler for general CIP
  • Highly effective in removing organic and inorganic scale deposits, thus improving operational efficiency
  • Highly effective in removing many coating across the pharmaceutical industry
  • Its defoaming properties make it suitable for use under conditions of high pressure and turbulence, improving cleaning performance
  • Very low phosphors and nitrogen content, which means that it meets most of the customers environmental requirements across all borders
  • Suitable for automatic dosing and control by conductivity, ensuring consistent delivery of product
Specifications
Volume (English) 5.3 gal.
Volume (Metric) 20 L