Learn More
Clifton™ Shaking Water Bath
Product Code. 11439539
Change view
Click to view available options
Capacity (Metric):
28 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11439539 28 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 11439539 Supplier Clifton Supplier No. NE528D
  • ONLINE EXCLUSIVE PRICE
    3336.79€
    2687.39€ / 1 pièces
    Économisez 649.40€
    19% de réduction
View all promotions
Expédition estimée: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Features precision PID control that provides simultaneous observation of set and actual temperature. Nickel Electro™ Clifton™ Shaking Water Bath with automatic tuning option that allows the bath to learn about your applications, environment and temperatures. Stainless steel recessed tray can be used for standing flasks on prior to and after immersion.
  • Precision PID control provides simultaneous observation of set and actual temperature.
  • Highly accurate PT100 sensor immersed directly in the bath.
  • Shaking system is quiet in use and by removing the trolley it can be used as a standard laboratory bath. The shaker trolley is highly stable in use, with a removable perforated shelf, unlike many traditional shaker baths where the whole shaker carriage has to be removed for loading and unloading.
  • Stainless steel recessed tray can be used for standing flasks on prior to and after immersion.
  • High quality corrosion resistant stainless steel tank, with a bright clean finish housed in a durable paint finished outer case.
  • Painted surfaces feature anti-bacterial finish, hygienic coating which actively inhibits bacterial growth, with good chemical resistance.
  • Heaters are concealed under the tank providing a clutter free working area easy to wipe clean. The heating footprint covers underside base of the tank, its low mass provides rapid energy transfer; quicker heat up times and a reduction in energy consumption with improved temperature uniformity.
  • LED displays measured temperatures in real-time, increments of ±0.1°C, using precision PID temperature control.
  • Settings retained even after turning the unit off.
  • Shaking control 0 to 400 strokes / 0 to 200 rpm - stroke length variable up to 40 mm (excluding NE5-10D which is fixed at 20 mm). Opening the side access port allows stroke length to be varied.
Specifications
Capacity (Metric) 28 L
Temperature Range 5°C to 99°C
Type Shaking Water Bath
Display LED
Model NE5-28 D
Temperature Resolution ±0.1°C
Temperature Uniformity ±0.1°C
Warranty 3 Years
Width (Metric) 715 mm
Width (Metric) Interior 500 mm
Height (Metric) 290 mm
Height (Metric) Interior 200 mm
Depth (Metric) Interior 300 mm
Electrical Requirements 230 V, 50/60 Hz, 1000 W
Voltage 230 V
Wattage 1000 W
Frequency 50/60 Hz