Learn More
STEMCELL Technologies ClonaCell™-HY Medium D without HAT
Product Code. 17168191
Encompass
Change view
Click to view available options
Quantity:
90 mL
Unit Size:
90mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17168191 90 mL 90mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17168191 Supplier STEMCELL Technologies Supplier No. 03810
 Restricted Item
Expédition estimée: 18-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Semi-solid methylcellulose-based hybridoma selection and cloning medium without HAT (serum-containing)
  • ClonaCell™‐HY Medium D without HAT is a semi-solid methylcellulose-based medium that does not contain any selection agents such as HAT (hypoxanthine, aminopterin, and thymidine), enabling alternative hybridoma or myeloma selection methods to be used
  • This product can be used to select and clone myelomas or hybridomas in one step by adding an appropriate selection agent to the medium
  • Individual parental clones and their progeny remain localized together in the semi-solid matrix as they grow to form distinct colonies
  • This prevents the loss of rare clones by overgrowth from faster-growing cells, as can occur during selection in a liquid medium, and facilitates the isolation of monoclonal colonies
  • The colonies can be easily picked from the semi-solid medium by manual or robotic methods and dispersed into a liquid medium for screening and expansion
Specifications
Cell Type Hybridomas
For Use With (Application) Cell Culture; Hybridoma Generation; Semi-Solid Cloning
Quantity 90 mL