Learn More
Skytec™ CMBH Boilersuit
Product Code. 31702839 Shop All Skytec Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Unit Size:
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31702839 X-Small Pack of 10
31702835 Small Pack of 10
31702836 Medium Pack of 10
31702823 Large Pack of 10
31702827 X-Large Pack of 10
31702832 2X-Large Pack of 10
31702821 3X-Large Pack of 10
31702825 4X-Large Pack of 10
31702822 5X-Large Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 31702839 Supplier Skytec™ Supplier No. CCCMB/001300/GN0
  • ONLINE EXCLUSIVE PRICE
    1860.43€
    1224.03€ / 10 pièces
    Économisez 636.40€
    34% de réduction
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Chemmaster garments offer strong resistance suited for a wide range of chemical handling tasks.
  • Ring-pull zip for easy donning and doffing when wearing gloves
  • Double storm-facing stud closure for increased protection
  • Helmet-compatible hood
  • Raglan sleeve for greater freedom of movement
  • Garment can be reused if not contaminated or damaged
  • Stitched and high-frequency welded seams
  • Tested to ASTM F903
Specifications
Color Green
Hood Type Attached
Material PVC
Size X-Small
Ankle Style Loose
Closure Type Zipper
Cuff Style Standard
Product Type Chemical Protective Clothing
Seam Type Welded