Learn More
behr Labor-Technik™ COD 24 Sample Reactor
Description
- Precision heating block thermostat
- For 24 reaction vessels
- Heating panel for particularly even temperature distribution over all sample positions
- Enclosure made of powder-coated steel and stainless steel.
- The elaborate heating block insulation ensures that housing temperatures stay within safe temperature ranges.
- Continuous grooves at the bottom of the boreholes simplify sample handling and increase user safety
- The grooves allow for pressure equalization when the sample vessels are inserted
- Even precisely fitting vessels can be smoothly inserted in the boreholes for user-friendly handling of the sample rack inserts
- Remaining fluid on the outside of the reaction vessel instantly evaporates when the samples are inserted in the heated block
- The grooves serve to discharge the vapor, thus preventing the breakage of glass due to glass vessel rupture
- In case of vessel overflow, the sample will drain off through the grooves, reducing the risk of acid damaging the heating block
Specifications
Specifications
| Description | Precision heating block thermostat for 24 reaction vessels RG, 20°C to 300°C |
| For Use With (Equipment) | TRS 300 Programmable Temperature and Time Controller |
| Type | COD 24 Sample Reactor |