Learn More
Code Corporation™ Code Reader™ 5200 Barcode Scanner with Age Verification
Product Code. 17983862 Shop All Brady Products
Change view
Click to view available options
Connection Type:
Cable
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connection Type Unit Size
17983862 Cable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17983862 Supplier Code Corporation™ Supplier No. 176696
  • ONLINE EXCLUSIVE PRICE
    565.00€
    447.40€ / 1 pièces
    Économisez 117.60€
    20% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Seamlessly connect your operations with the CR5200. The 5200 Barcode Scanner reads barcodes in any orientation, offers hands-free operation and can verify age by scanning ID cards.
  • Get high-speed, omnidirectional reading of 1D and 2D barcode symbologies
  • Features age verification when scanning ID cards
  • Quickly read barcodes on mobile device screens
  • Compact and adjustable for any reading direction
  • Provides user feedback with LED and audible tone
  • Features extended read ranges
Specifications
Height (Metric) 139.7 mm
Height (English) 5.5 in.
Width (Metric) 76.2 mm
Width (English) 3 in.
Depth (Metric) 88.9 mm
Depth (English) 3.5 in.
Weight (Metric) 318 g
Weight (English) 11.2 oz
Field View 51° Horizontal by 39.4° Vertical
Includes CR5200, USB Cable
Type Barcode Scanner
Connection Type Cable
Data Memory 1 MB NOR Flash, 16 MB RAM
Power Supply USB
Resolution 1280 x 960
Alternate Part Number CR5215-200-C500-C286