Learn More
Abcam Coenzyme A Assay Kit (Fluorometric - Green)
Product Code. 32118271 Shop All Abcam Products
Change view
Click to view available options
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
32118271 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32118271 Supplier Abcam Supplier No. ab138889200Test
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Measure coenzyme A in 1h in Cell/Tissue Extracts with Coenzyme A Assay Kit AB138889. Sensitivity = 40 nM. Cited in 8 publications. Use with Microplate reader.
Measure coenzyme A in 1h in Cell/Tissue Extracts with Coenzyme A Assay Kit AB138889. Sensitivity = 40 nM. Cited in 8 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 200 Tests
Regulatory Status RUO