Learn More
Pacific Handy Cutter Coil Lanyard with Swivel Connector
Product Code. 17779047
Change view
Click to view available options
Length (English):
12.9 in.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
17779047 12.9 in. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17779047 Supplier Pacific Handy Cutter Supplier No. CL36
  • ONLINE EXCLUSIVE PRICE
    214.07€
    185.29€ / 50 pièces
    Économisez 28.78€
    13% de réduction
Expédition estimée: 24-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The CL-36 Coil Lanyard is multipurpose and the elastic cord easily stretches to facilitate any normal cutting motions.
  • Users observe noticeable increases in cutter retention and report substantial reductions in replacement costs when utilized in combination with any PHC safety cutter and holster.
  • It can also be attached to a work station to keep the tool close at hand.
Specifications
Product Type Knife Lanyard
For Use With (Equipment) All Cutters
Length (English) 12.9 in.
Length (Metric) 330 mm
Sterility Non-sterile