Learn More
KGW Isotherm™ Cold Trap
Product Code. 10264892
Change view
Click to view available options
Model:
KF 29-GL
KF 29-GL-A
KFL 29-GL
KFL 29-GL-A
Includes:
12C Dewar
12C Dewar, Spout
18C Dewar
18C Dewar, Spout
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Includes Unit Size
10264892 KF 29-GL-A 12C Dewar, Spout Each
10475602 KFL 29-GL-A 18C Dewar, Spout Each
10202022 KF 29-GL 12C Dewar Each
10789444 KFL 29-GL 18C Dewar Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10264892 Supplier KGW Isotherm™ Supplier No. 17372
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
Expédition estimée: 17-12-2026
Add to basket
Add to basket
Specifications
Capacity (Metric) 150 mL (Condensate), 1 L (Coolant)
Includes 12C Dewar, Spout
Quantity 1 Pack
Type Cold Trap (Short)
Model KF 29-GL-A
Connection Type Glass Thread GL18 with Olive