Learn More
STEMCELL Technologies Collagen Solution
Product Code. 15212755
Encompass_Preferred
Change view
Click to view available options
Quantity:
35 mL
Unit Size:
35mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15212755 35 mL 35mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15212755 Supplier STEMCELL Technologies Supplier No. 04902
 Restricted Item
Expédition estimée: 20-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For preparation of collagen gels and for coating cell culture surfaces
  • MegaCult™-C Complete Kit with Cytokines includes all reagents required for the optimal growth of megakaryocyte progenitors (CFU-Mk) within double chamber slides and for the detection of CFU-Mk colonies by immunocytochemical staining
  • MegaCult™-C medium is optimized for the growth of CFU-Mk in human bone marrow, mobilized peripheral blood and cord blood samples
  • It is suitable for use with CD34+-enriched cells, mononuclear cells and cells isolated by other purification methods
Specifications
For Use With (Application) Cell Culture, Colony Assay, Functional Assay
Quantity 35 mL
Cell Type Hematopoietic Stem and Progenitor Cells