Learn More
Conductivity Calibration Standards, YSI™
Product Code. 12766945
Change view
Click to view available options
Quantity:
1 qt.
8 x 1 pt.
Packaging:
Glass Bottle
Plastic Bottle
Conductivity:
1,000μS/cm (Fresh water)
10,000μS/cm
50,000μS/cm (sea water)
Unit Size:
Each
Pack of 8
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Conductivity Unit Size
12766945 8 x 1 pt. Plastic Bottle 10,000μS/cm Pack of 8
12736945 1 qt. Glass Bottle 10,000μS/cm Each
12756945 8 x 1 pt. Plastic Bottle 1,000μS/cm (Fresh water) Pack of 8
12776945 8 x 1 pt. Plastic Bottle 50,000μS/cm (sea water) Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12766945 Supplier YSI Yellow Spring Instrument Supplier No. 060911
  • ONLINE EXCLUSIVE PRICE
    155.02€
    147.74€ / 8 pièces
    Économisez 7.28€
    4% de réduction
 Restricted Item
Expédition estimée: 15-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
NIST-traceable conductivity solutions to calibrate both field instruments and lab instruments and cells together for maximum accuracy
  • Conductivity solutions are inspected with reference to primary standard solutions according to OIML recommendation 56
  • Bottles include a table of corrections at temperatures between 20 and 30°C
  • Available as: 1 Quart or in a box of 8 individual pints
TRUSTED_SUSTAINABILITY
Specifications
Packaging Plastic Bottle
Conductivity 10,000μS/cm
Type Conductivity Calibration Standard
Quantity 8 x 1 pt.