Learn More
Consort™ Multi-parameter Analyzer Kits
Product Code. 12157810
Change view
Click to view available options
Type:
Conductivity Kit
pH/Conductivity/DO kit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12157810 Conductivity Kit Each
11965458 pH/Conductivity/DO kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12157810 Supplier Consort™ Supplier No. C6010K
  • ONLINE EXCLUSIVE PRICE
    1309.95€
    1011.00€ / 1 pièces
    Économisez 298.95€
    22% de réduction
Expédition estimée: 13-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Includes all the components needed for electrochemical measurement. Consort™ Multi-parameter Analyzer Kits include a high-quality, portable meter as well as the necessary electrodes; buffers, solutions and standards; and adapters and accessories required for field and laboratory use.
C5020T: pH/EC/DO Meter Kit
The C5020 is a complete and accurate portable meter that can measure all common electrochemical parameters such as pH, ORP, conductivity, and dissolved oxygen, plus salinity and TDS. It is an all-round instrument with a large conductivity range and measurement storing capabilities. The large graphical display makes it easy to read the measurement, and the stability indicator/algorithm ensures steady, accurate readings.
  • C5020 Multi-Parameter Analyzer
  • Electrodes: pH/ATC electrode SP10T, Conductivity/ATC electrode SK10T, Dissolved oxygen electrode SZ10T
  • 2 x 50mL pH buffers (pH 4 and 7)
  • 50mL Electrolyte solution (3M KCl)
  • 50mL conductivity standard (0.01M KCl)
  • Mains adaptor (100–240 VAC, EU/US)
C6010K: EC Meter Kit
The C6010 is a single channel multi-parameter analyzer suitable as a bench-top or portable device. A corrosion-resistant gold-plated BNC connector ensures a long life and accepts different electrodes. An easy-to-use menu and built-in help system enables users to configure and utilize the instrument successfully. Besides the pre-programmed pH buffers and EC standards, other tables can be added, including the complete temperature table.
  • C6010 Multi-Parameter Analyzer
  • Glass conductivity/ATC electrode SK20T
  • 50mL conductivity standard (0.01M KCl)
  • Carrying case
  • Mains adaptor (100–240 VAC, EU/US)
Specifications
Accuracy (Conductivity) 0.5%
Accuracy (Temperature) 0.1°C
Calibration GLP
Type Conductivity Kit
Dimensions (L x W x H) 12 x 25 x 5 cm
Display Type LCD
Frequency 50/60Hz
Inputs BNC (measurement) and Banana for Pt1000 (temperature)
Includes C6010 Meter, conductivity/ATC electrode SK20T, conductivity standard, carrying case, mains adapter
Model C6010K
Probe Type Conductivity/ATC glass electrode SK20T
Range (Conductivity) 0 to 2000 mS/cm
Range (Resistivity) 0 to 200 Mohm/cm
Range (Salinity) 0 to 70
Range (TDS) 0 to 100.0 g/L
Resolution (Conductivity) 0.001 μS/cm
Resolution (pH) 0.001 pH
Resolution (Resistivity) 1 ohm/cm
Resolution (Temperature) 0.1°C
Resolution (TDS) 0.01 mg/L
Temperature (Metric) Range -5°C to +105°C
Voltage 100/240VAC
Weight (Metric) 600 g