Learn More
Conductivity Solution Standard, Hanna Instruments
Product Code. 10462521
Change view
Click to view available options
Quantity:
460 mL
Unit Size:
460mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10462521 460 mL 460mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10462521 Supplier Hanna Instruments Supplier No. HI70300L
 Restricted Item
Expédition estimée: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A storage solution prepared with reagent grade chemicals that can be used to ensure optimum performance of your pH and ORP electrodes.
  • Storage solution prepared with reagent grade chemicals that can be used to ensure optimum performance of your pH and ORP electrodes
  • Special formulation to minimize microbial growth and osmotic/diffusion effects between the solution and inner reference electrolyte
  • Manufactured specifically for pH and ORP electrodes
  • Clearly marked expiration date and lot number
Specifications
Chemical Name or Material Conductivity Solution Standard
Quantity 460 mL