missing translation for 'onlineSavingsMsg'
Learn More

Liofilchem™ Contact Slide 1

Product Code. 11954369
Encompass_Preferred
Change view
Click to view available options
Quantity:
120 Slides
20 Slides
Unit Size:
Pack of 120
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11954369 20 Slides Pack of 20
11944339 120 Slides Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11954369 Supplier Liofilchem™ Supplier No. 525262

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PCA and TTC and Neutralizing / VRBG Agar and Neutralizing, Flex Dip-slide with a selective medium for detection of Enterobacteriaceae and a non selective medium for total bacterial count.

  • Contact Slide 1 is a ready-to-use device with two different media coated onto a plastic support used for the microbial monitoring of surfaces and liquids even in the presence of residues of disinfectants. The selective medium allows the isolation and enumeration of Enterobacteriaceae . The other medium is used to obtain the total bacterial count.

Specifications

Certifications/Compliance ISO 18593:2004
Content And Storage 10°C to 25°C
Detects Bacterial Count, Enterobacteriaceae
Format Slide
Manufacturing Origin Italy
Product Type Contact Slide
Sterility Non-sterile
Target Organism Class Coliforms, Gram-positive Organisms, Coliforms
Formulation PCA, TTC, Neutralizing/VRBG Agar, Neutralizing
For Use With (Application) For Bacterial Count and Enterobacteriaceae Detection
Incubation Conditions At 36 ± 1°C for 24 to 48 hr.
Incubation Time 24 to 48hr.
Quantity 20 Slides
Regulatory Status ISO 18593:2004
Slide Type Flex
Species Escherichia coli, Proteus species, Klebsiella species, Pseudomonas species
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.