Learn More
Invitrogen™ CRISPR TrueGuide™ on-line tool
Description
Invitrogen TrueGuide Synthetic gRNAs are ready-to-use, synthetic gRNAs designed to provide specific and high-efficiency knockout of your target gene.
TrueGuide Synthetic gRNAs both maximize and simplify the performance of genome editing experiments so researchers can create more relevant cellular models and make new discoveries. Custom TrueGuide Synthetic gRNAs are available in two package sizes: 1.5 nmol and 3 nmol.
Spécification
Spécification
| Product Type | Synthetic gRNA |
| Content And Storage | 1 Dried RNA Oligo, Store at Room Temperature |
| Quantity | 1.5 nmol |
| Product Line | TrueGuide |
For Research Use Only. Not for use in diagnostic procedures.