Learn More
Welch™ CRVpro™ Starter Package
Product Code. 15721918 Shop All Welch Ilmvac Products
Change view
Click to view available options
Model:
CRVpro 4
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
15721918 CRVpro 4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15721918 Supplier Welch™ Supplier No. 330071
  • ONLINE EXCLUSIVE PRICE
    2991.15€
    2865.00€ / 1 pièces
    Économisez 126.15€
    4% de réduction
View all promotions
Expédition estimée: 01-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
The robust vacuum pump CRVpro belongs to the new high-performance two-stage rotary vane pumps generation of Welch.
  • CRVpro Starter Package Includes:
    • Oil mist separator AKD 16
    • 2-way ball valve
    • Hose connector DN 16KF-10/8 mm
    • Vacuum hose (1.5 m)
    • Oil change set
TRUSTED_SUSTAINABILITY
Specifications
Includes CRVpro with Oil Mist Separator AKD 16, 2-Way Ball Valve, Hose Connector, Vacuum Hose (1.5 m), Oil Change Set
Model CRVpro 4
Pump Type Rotary Vane Vacuum Pump